Assay Details
| Gene Symbol: ROM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174174.3 | NP_776599.2 | 2 - 3 | 1045 | 105 | |
| GenBank mRNA | BC118152.1 | - | 2 - 3 | 1045 | 105 | |
| D83385.1 | - | 2 - 3 | 855 | 105 | ||
| U72027.1 | - | 2 - 3 | 840 | 105 | ||
More Information
Set Membership: |
|

ROM1
Genomic Map