Assay Details
| Gene Symbol: IMPA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174361.4 | NP_776786.1 | 5 - 6 | 404 | 75 | |
| GenBank mRNA | BC118411.1 | - | 5 - 6 | 405 | 75 | |
| BT030590.2 | - | 5 - 6 | 370 | 75 | ||
| J05394.1 | - | 5 - 6 | 414 | 75 | ||
More Information
Set Membership: |
|

IMPA1
Genomic Map