Assay Details
| Gene Symbol: THTPA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174470.4 | NP_776895.1 | 2 - 3 | 222 | 88 | |
| GenBank mRNA | BC105468.1 | - | 2 - 3 | 223 | 88 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC105468.1, NM_174470.4 transcripts.
Set Membership: |
|

THTPA
Genomic Map