Assay Details
| Gene Symbol: CYP3A5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174531.3 | NP_776956.2 | 9 - 10 | 951 | 125 | |
| GenBank mRNA | BC114003.1 | - | 9 - 10 | 952 | 125 | |
| Y10214.1 | - | 870 | 125 | |||
More Information
Set Membership: |
|

CYP3A5
Genomic Map