Assay Details
| Gene Symbol: PRKAG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174586.2 | NP_777011.2 | 1 - 2 | 54 | 109 | |
| GenBank mRNA | BC113296.1 | - | 1 - 2 | 54 | 109 | |
| BT025456.1 | - | 1 - 2 | 57 | 109 | ||
More Information
Set Membership: |
|

PRKAG1
Genomic Map