Assay Details
| Gene Symbol: BGN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178318.4 | NP_847888.2 | 4 - 5 | 671 | 66 | |
| GenBank mRNA | BC118460.1 | - | 4 - 5 | 672 | 66 | |
| BT021201.1 | - | 4 - 5 | 649 | 66 | ||
| L07953.1 | - | 4 - 5 | 66 | |||
More Information
Set Membership: |
|

BGN
Genomic Map