Assay Details
| Gene Symbol: G6PC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183364.3 | NP_899208.2 | 1 - 2 | 256 | 60 | |
| GenBank mRNA | AY279358.1 | - | 1 - 2 | 272 | 60 | |
| BC118357.1 | - | 1 - 2 | 257 | 60 | ||
More Information
Set Membership: |
|

G6PC3
Genomic Map