Assay Details
| Gene Symbol: CCL11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205773.2 | NP_991342.1 | 1 - 2 | 173 | 91 | |
| GenBank mRNA | AJ132003.2 | - | 1 - 2 | 144 | 91 | |
| BC114735.1 | - | 1 - 2 | 173 | 91 | ||
| EU744565.1 | - | 1 - 2 | 123 | 91 | ||
More Information
Set Membership: |
|

CCL11
Genomic Map