Assay Details
| Gene Symbol: HCRTR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001048182.1 | NP_001041647.1 | 3 - 4 | 625 | 74 | |
| GenBank mRNA | AB092488.1 | - | 1 - 2 | 121 | 74 | |
| BC149920.1 | - | 5 - 6 | 1329 | 74 | ||
| DQ874350.1 | - | 3 - 4 | 625 | 74 | ||
More Information
Set Membership: |
|

HCRTR1
Genomic Map