Assay Details
| Gene Symbol: PTPRN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075131.1 | NP_001068599.1 | 7 - 8 | 1177 | 59 | |
| GenBank mRNA | AF075170.1 | - | 7 - 8 | 1138 | 59 | |
| BC119839.1 | - | 7 - 8 | 1177 | 59 | ||
More Information
Set Membership: |
|

PTPRN
Genomic Map