Assay Details
| Gene Symbol: HSPA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075148.1 | NP_001068616.1 | 3 - 4 | 475 | 84 | |
| GenBank mRNA | BC119953.1 | - | 3 - 4 | 475 | 84 | |
| BT030726.1 | - | 3 - 4 | 500 | 84 | ||
More Information
Set Membership: |
|

HSPA5
Genomic Map