Assay Details
| Gene Symbol: HEXA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075164.2 | NP_001068632.1 | 13 - 14 | 1556 | 113 | |
| GenBank mRNA | BC112560.1 | - | 13 - 14 | 1539 | 113 | |
| BT026152.1 | - | 13 - 14 | 1556 | 113 | ||
More Information
Set Membership: |
|

HEXA
Genomic Map