Assay Details
| Gene Symbol: LARP7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075184.2 | NP_001068652.2 | 12 - 13 | 1766 | 92 | |
| GenBank mRNA | BC119943.1 | - | 6 - 7 | 1006 | 92 | |
| BC122741.1 | - | 2 - 3 | 382 | 92 | ||
More Information
Set Membership: |
|

LARP7
Genomic Map