Assay Details
| Gene Symbol: METTL2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075246.2 | NP_001068714.1 | 2 - 3 | 209 | 153 | |
| GenBank mRNA | BC120275.1 | - | 2 - 3 | 211 | 153 | |
More Information
Set Membership: |
|

METTL2A
Genomic Map