Assay Details
| Gene Symbol: SIKE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075281.2 | NP_001068749.1 | 3 - 4 | 507 | 77 | |
| GenBank mRNA | BC120197.1 | - | 3 - 4 | 508 | 77 | |
More Information
Set Membership: |
|

SIKE1
Genomic Map