Assay Details
| Gene Symbol: IL33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075297.1 | NP_001068765.1 | 8 - 9 | 826 | 65 | |
| GenBank mRNA | BC123562.1 | - | 8 - 9 | 826 | 65 | |
More Information
Set Membership: |
|

IL33
Genomic Map