Assay Details
| Gene Symbol: UFD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075300.1 | NP_001068768.1 | 1 - 2 | 36 | 65 | |
| GenBank mRNA | BC120441.1 | - | 1 - 2 | 36 | 65 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC120441.1, NM_001075300.1 transcripts.
Set Membership: |
|

UFD1
Genomic Map