Assay Details
| Gene Symbol: UFD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075300.1 | NP_001068768.1 | 5 - 6 | 454 | 70 | |
| GenBank mRNA | BC120441.1 | - | 5 - 6 | 454 | 70 | |
More Information
Set Membership: |
|

UFD1
Genomic Map