Assay Details
| Gene Symbol: MFSD10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075304.1 | NP_001068772.1 | 12 - 13 | 1454 | 67 | |
| GenBank mRNA | BC119845.1 | - | 12 - 13 | 1454 | 67 | |
More Information
Set Membership: |
|

MFSD10
Genomic Map