Assay Details
| Gene Symbol: ANP32E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075306.1 | NP_001068774.1 | 3 - 4 | 625 | 139 | |
| GenBank mRNA | BC120174.1 | - | 3 - 4 | 625 | 139 | |
More Information
Set Membership: |
|

ANP32E
Genomic Map