Assay Details
| Gene Symbol: SEPHS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075316.1 | NP_001068784.1 | 7 - 8 | 1028 | 61 | |
| GenBank mRNA | BC120303.1 | - | 7 - 8 | 1028 | 61 | |
More Information
Set Membership: |
|

SEPHS1
Genomic Map