Assay Details
| Gene Symbol: TOMM70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075328.1 | NP_001068796.1 | 11 - 12 | 1750 | 132 | |
| GenBank mRNA | BC123444.1 | - | 11 - 12 | 1750 | 132 | |
| HM347499.1 | - | 11 - 12 | 1675 | 132 | ||
More Information
Set Membership: |
|

TOMM70
Genomic Map