Assay Details
| Gene Symbol: TOMM70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075328.1 | NP_001068796.1 | 1 - 2 | 406 | 58 | |
| GenBank mRNA | BC123444.1 | - | 1 - 2 | 406 | 58 | |
| HM347499.1 | - | 1 - 2 | 331 | 58 | ||
More Information
Set Membership: |
|

TOMM70
Genomic Map