Assay Details
Gene Symbol: TOMM70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075328.1 | NP_001068796.1 | 2 - 3 | 576 | 81 | |
GenBank mRNA | BC123444.1 | - | 2 - 3 | 576 | 81 | |
HM347499.1 | - | 2 - 3 | 501 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
