Assay Details
| Gene Symbol: TOMM70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075328.1 | NP_001068796.1 | 3 - 4 | 709 | 93 | |
| GenBank mRNA | BC123444.1 | - | 3 - 4 | 709 | 93 | |
| HM347499.1 | - | 3 - 4 | 634 | 93 | ||
More Information
Set Membership: |
|

TOMM70
Genomic Map