Assay Details
| Gene Symbol: KYAT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075370.1 | NP_001068838.1 | 10 - 11 | 994 | 124 | |
| GenBank mRNA | BC120067.1 | - | 10 - 11 | 994 | 124 | |
More Information
Set Membership: |
|

KYAT3
Genomic Map