Assay Details
| Gene Symbol: TP53I3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075378.1 | NP_001068846.1 | 2 - 3 | 511 | 71 | |
| GenBank mRNA | BC120317.1 | - | 2 - 3 | 511 | 71 | |
More Information
Set Membership: |
|

TP53I3
Genomic Map