Assay Details
| Gene Symbol: ANKMY2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075384.2 | NP_001068852.1 | 7 - 8 | 1086 | 66 | |
| GenBank mRNA | BC120022.1 | - | 7 - 8 | 1087 | 66 | |
More Information
Set Membership: |
|

ANKMY2
Genomic Map