Assay Details
| Gene Symbol: ANKRD37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075392.1 | NP_001068860.1 | 2 - 3 | 297 | 55 | |
| GenBank mRNA | BC120289.1 | - | 2 - 3 | 297 | 55 | |
More Information
Set Membership: |
|

ANKRD37
Genomic Map