Assay Details
| Gene Symbol: ANKRD37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075392.1 | NP_001068860.1 | 3 - 4 | 381 | 75 | |
| GenBank mRNA | BC120289.1 | - | 3 - 4 | 381 | 75 | |
More Information
Set Membership: |
|

ANKRD37
Genomic Map