Assay Details
| Gene Symbol: MALSU1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075398.2 | NP_001068866.1 | 3 - 4 | 528 | 73 | |
| GenBank mRNA | BC120471.1 | - | 3 - 4 | 530 | 73 | |
More Information
Set Membership: |
|

MALSU1
Genomic Map