Assay Details
| Gene Symbol: HDDC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075401.1 | NP_001068869.1 | 5 - 6 | 548 | 63 | |
| GenBank mRNA | BC120449.1 | - | 5 - 6 | 548 | 63 | |
More Information
Set Membership: |
|

HDDC2
Genomic Map