Assay Details
| Gene Symbol: SPX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075407.1 | NP_001068875.1 | 5 - 6 | 360 | 89 | |
| GenBank mRNA | BC120360.1 | - | 5 - 6 | 360 | 89 | |
More Information
Set Membership: |
|

SPX
Genomic Map