Assay Details
| Gene Symbol: LIN7C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075431.2 | NP_001068899.1 | 3 - 4 | 264 | 61 | |
| GenBank mRNA | BC120124.1 | - | 3 - 4 | 265 | 61 | |
More Information
Set Membership: |
|

LIN7C
Genomic Map