Assay Details
| Gene Symbol: ALG6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075432.3 | NP_001068900.2 | 8 - 9 | 914 | 110 | |
| GenBank mRNA | BC120396.1 | - | 8 - 9 | 914 | 110 | |
More Information
Set Membership: |
|

ALG6
Genomic Map