Assay Details
| Gene Symbol: CNPY3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075439.2 | NP_001068907.1 | 3 - 4 | 425 | 57 | |
| GenBank mRNA | BC119829.1 | - | 3 - 4 | 426 | 57 | |
More Information
Set Membership: |
|

CNPY3
Genomic Map