Assay Details
| Gene Symbol: USB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075477.2 | NP_001068945.1 | 2 - 3 | 320 | 76 | |
| GenBank mRNA | BC122807.1 | - | 2 - 3 | 321 | 76 | |
More Information
Set Membership: |
|

USB1
Genomic Map