Assay Details
| Gene Symbol: NMNAT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075486.1 | NP_001068954.1 | 7 - 8 | 690 | 53 | |
| GenBank mRNA | BC120344.1 | - | 7 - 8 | 690 | 53 | |
More Information
Set Membership: |
|

NMNAT2
Genomic Map