Assay Details
| Gene Symbol: ZW10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075489.1 | NP_001068957.1 | 14 - 15 | 2023 | 72 | |
| GenBank mRNA | BC119883.1 | - | 14 - 15 | 2023 | 72 | |
More Information
Set Membership: |
|

ZW10
Genomic Map