Assay Details
| Gene Symbol: AGGF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075532.2 | NP_001069000.2 | 3 - 4 | 856 | 72 | |
| GenBank mRNA | BC119837.1 | - | 3 - 4 | 830 | 72 | |
More Information
Set Membership: |
|

AGGF1
Genomic Map