Assay Details
| Gene Symbol: YRDC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075550.1 | NP_001069018.1 | 4 - 5 | 860 | 64 | |
| GenBank mRNA | BC120307.1 | - | 4 - 5 | 860 | 64 | |
More Information
Set Membership: |
|

YRDC
Genomic Map