Assay Details
| Gene Symbol: ERGIC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075563.1 | NP_001069031.1 | 9 - 10 | 717 | 83 | |
| GenBank mRNA | BC122615.1 | - | 9 - 10 | 717 | 83 | |
More Information
Set Membership: |
|

ERGIC2
Genomic Map