Assay Details
| Gene Symbol: KCTD15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075568.1 | NP_001069036.1 | 5 - 6 | 658 | 58 | |
| GenBank mRNA | BC119910.1 | - | 5 - 6 | 658 | 58 | |
| BT029851.1 | - | 5 - 6 | 606 | 58 | ||
More Information
Set Membership: |
|

KCTD15
Genomic Map