Assay Details
| Gene Symbol: SRSF11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075621.1 | NP_001069089.1 | 2 - 3 | 246 | 55 | |
| GenBank mRNA | BC118154.1 | - | 2 - 3 | 246 | 55 | |
More Information
Set Membership: |
|

SRSF11
Genomic Map