Assay Details
| Gene Symbol: PWWP3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075622.1 | NP_001069090.1 | 11 - 12 | 1851 | 68 | |
| GenBank mRNA | BC114659.1 | - | 8 - 9 | 1274 | 68 | |
| BC123693.1 | - | 11 - 12 | 1851 | 68 | ||
More Information
Set Membership: |
|

PWWP3A
Genomic Map