Assay Details
| Gene Symbol: PWWP3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075622.1 | NP_001069090.1 | 1 - 2 | 125 | 152 | |
| GenBank mRNA | BC123693.1 | - | 1 - 2 | 125 | 152 | |
More Information
Set Membership: |
|

PWWP3A
Genomic Map