Assay Details
| Gene Symbol: DCTN6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075638.2 | NP_001069106.1 | 6 - 7 | 489 | 57 | |
| GenBank mRNA | BC118351.1 | - | 6 - 7 | 490 | 57 | |
More Information
Set Membership: |
|

DCTN6
Genomic Map