Assay Details
| Gene Symbol: ATP6V1G2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075664.1 | NP_001069132.1 | 1 - 2 | 157 | 62 | |
| GenBank mRNA | BC111675.1 | - | 1 - 2 | 140 | 62 | |
| BC119976.1 | - | 1 - 2 | 157 | 62 | ||
More Information
Set Membership: |
|

ATP6V1G2
Genomic Map