Assay Details
| Gene Symbol: IRAK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075696.2 | NP_001069164.1 | 1 - 2 | 136 | 82 | |
| GenBank mRNA | BC120001.1 | - | 1 - 2 | 137 | 82 | |
| EU528620.1 | - | 1 - 2 | 131 | 82 | ||
| EU528621.1 | - | 1 - 2 | 131 | 82 | ||
More Information
Set Membership: |
|

IRAK2
Genomic Map