Assay Details
| Gene Symbol: RAP2C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075700.1 | NP_001069168.1 | 2 - 3 | 685 | 117 | |
| GenBank mRNA | BC123730.1 | - | 2 - 3 | 685 | 117 | |
More Information
Set Membership: |
|

RAP2C
Genomic Map